Bis(dibenzothiazepinyl) Piperazine, Pharmaceutical Analytical Impurities
Katalog # 1A00150.
Leverantör: USP
CAS-nummer:
Specifikationer
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:945668-94-0
- Beskrivning:Bis(dibenzothiazepinyl) Piperazine
- Synonym(s):1,4-Bis(dibenzo[b,f][1,4]thiazepin-11-yl)piperazine
- Benämning:Bis(dibenzothiazepinyl) Piperazine
Specifikationer
Om denna produkt
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.