Order Entry
Sweden
ContactUsLinkComponent
Azacitidine Isomer -3, Pharmaceutical Analytical Impurities
Azacitidine Isomer -3, Pharmaceutical Analytical Impurities
Katalog # 1A00270.
Supplier:  USP
CAS Number:  
Azacitidine Isomer -3, Pharmaceutical Analytical Impurities
Katalog # 1A00270.
Supplier:  USP
CAS Number:  

Specifications

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • Beskrivning:
    Azacitidine Isomer -3
  • Synonym(s):
    1-β-D-Ribofuranosyl-3-aminocarbonyl guanidine acetate
  • Benämning:
    Azacitidine Isomer -3

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.