ContactUsLinkComponent
Amlodipine Analog, Pharmaceutical Analytical Impurities
Amlodipine Analog, Pharmaceutical Analytical Impurities
Leverantör:  USP
Certifikat
Amlodipine Analog, Pharmaceutical Analytical Impurities
Leverantör:  USP
Specifikationer
  • CAS-nr:
    400024-11-5
  • Synonym(s):
    dimethyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, maleate
  • Benämning:
    Amlodipine Methyl Analog
Specifikationer