6-Methyl-4-phenylchromanol, Pharmaceutical Analytical Impurities
Katalog # 1A01050.
Leverantör: USP
CAS-nummer:
Specifikationer
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:209747-04-6
- Beskrivning:6-Methyl-4-phenylchromanol
- Synonym(s):6-Methyl-4-phenylchroman-2-ol
- Benämning:6-Methyl-4-phenylchromanol
Specifikationer
Om denna produkt
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.