Order Entry
Sweden
ContactUsLinkComponent
Hydroxy Ethinyl Estradiol, Pharmaceutical Analytical Impurities
Hydroxy Ethinyl Estradiol, Pharmaceutical Analytical Impurities
Katalog # 1A00110.
Leverantör:  USP
CAS-nummer:  
Hydroxy Ethinyl Estradiol, Pharmaceutical Analytical Impurities
Katalog # 1A00110.
Leverantör:  USP
CAS-nummer:  

Specifikationer

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • CAS-nr:
    56324-28-8
  • Beskrivning:
    6Beta-Hydroxy Ethinyl Estradiol
  • Synonym(s):
    (6R,8R,9S,13S,14S,17R)-17-ethynyl-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,6,17-triol
  • Benämning:
    6Beta-Hydroxy Ethinyl Estradiol

Specifikationer

Om denna produkt

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.