6-alpha-Hydroxybudesonide, Pharmaceutical Analytical Impurities
Katalog # 1A01390.
Leverantör: USP
CAS-nummer:
Specifikationer
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:577777-51-6
- Beskrivning:6alpha-Hydroxybudesonide
- Synonym(s):(2S,6aR,6bS,7S,8aS,8bS,11aR,12aS,12bS)-2,7-Dihydroxy-8b-(2-hydroxyacetyl)-6a,8a-dimethyl-10-propyl-1,2,6a,6b,7,8,8a,8b,11a,12,12a,12b-dodecahydro-4H-naphtho[2',1':4,5]indeno[1,2-d][1,3]dioxol-4-one
- Benämning:6alpha-Hydroxybudesonide
Specifikationer
Om denna produkt
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.