5-Dimethylaminobutyryl Citalopram Oxalate, Pharmaceutical Analytical Impurities
Katalog # 1A01770.
Supplier: USP
CAS Number:
Specifications
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- Beskrivning:5-Dimethylaminobutyryl Citalopram Oxalate
- Synonym(s):4-(dimethylamino)-1-(1-(3-(dimethylamino)propyl)-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-yl)butan-1-one, oxalate
- Benämning:5-Dimethylaminobutyryl Citalopram Oxalate
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.