2,3-Dimethyl-2,3-butanediamine Dihydrochloride, Pharmaceutical Analytical Impurities
Katalog # 1A01340.
Leverantör: USP
CAS-nummer:
Specifikationer
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:75804-28-3
- Beskrivning:2,3-Dimethyl-2,3-butanediamine Dihydrochloride
- Synonym(s):2,3-dimethylbutane-2,3-diamine, hydrochloride
- Benämning:2,3-Dimethyl-2,3-butanediamine Dihydrochloride
Specifikationer
Om denna produkt
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.