Specifications
- Steril:-
- Typ:With attached cap
- Kapacitet:1,5 ml
- Färg:Genomskinlig
- Material:PP
- Seq:03
- Förp.:500
- Max g-tal:20000×g
Specifications
About this item
The disposable microtubes with 1,5 ml nominal volume are made from high-quality PP. The quality level BIO-CERT® PCR QUALITY ensures the required cleanliness for PCR-setups, storage of PCR samples and other applications with high cleanliness requirements. This includes absence of DNA, RNase, DNase, pyrogenes and PCR inhibitors.
- High cleanliness for demanding applications
- Tight sealing snap lid
- Can be opened and closed with one hand
They are available with an easy-to-open snap-on lid in versions with or without lid locking. Lid locking prevents unintended opening of the lid when heating the samples inside the microtubes. These disposable microtubes can be centrifuged up to the maximum RCF value stated and autoclaved at 121 °C (2 bar) according to DIN EN 285. For a reading of the approximate volume, they have an easy-to-read graduation. For easy identification of samples, different colours are available.