Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Centrifuger
Rotorer
Rotors, Avanti JA 12
Rotors, Avanti JA 12
Rotors, Avanti JA 12
Supplier:
Beckman Coulter
Certificates
Expand image
Rotors, Avanti JA 12
Supplier:
Beckman Coulter
About this item
Specifications
Catalog No:
521-3944
521-3945
Kapacitet:
600 ml
600 ml
Radie:
144 mm
144 mm
Max. RPM:
12000
12000
Specifications