
KMUH (N-(κ-maleimidoundecanoic acid) hydrazide trifluoroacetic acid salt), Pierce™
Catalog # PIER22111
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Förp.:50 mg
- Seq:2370
Specifications
About this item
:
:409.40 g/mol
:GHS07
Storage Temperature:Kylskåp
CAS Number:1593661-78-9
:359436-62-7
Formula:C₁₅H₂₅N₃O₃·CF₃CO₂H