Anti-IgG gamma goat polyclonal antibody (AP (Alkaline phosphatase))
KPLI5220-0348
- Antikroppstyp:Sekundär
- Antikroppsnamn:Immunoglobulin G gamma chain
- Antigensymbol:IgG gamma
- Klonalitet:Polyclonal
- Konjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flödescytometri:Yes
- Värd:Goat
- ImmunKemi:Yes
- Immunutfällning:Yes
- Reaktivitet:Human
- Western blot:Yes
- Epitop:γ chain
- Förp.:1 mg
Secondary Goat Anti-IgG(gamma) reacts with human
Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Human