Specifications
- Beskrivning:E-A-Rcaps™ banded earplugs, 23 dB
- Förp.:400 PAIR
Specifications
About this item
The E-A-Rcaps™ banded hearing protector is fitted with semi-aural eartips designed to seal the entrance of the ear canal to help reduce exposure to hazardous levels of noise.
- Reduces noise levels up to 23 dB
- Flexible and durable band can be washed and reused multiple times
- Soft foam pod shaped caps to block the ear canal
- Ideal for intermittent wear, the band can be easily stored around the neck when not in use
- Compatible with the 3M™ E-A-Rfit™ Dual-Ear validation system
Get one of the lightest banded hearing protectors on the market with 3M™ E-A-Rcaps™ Banded Earplugs. They feature comfortable foam caps on a flexible band. The soft and smooth polyurethane cap shaped earplug effectively seals the ear canal, providing 23dB CE certified protection, tested to EN 352.
Ideal for dirty environments, the pods are simple to push in without the need for rolling down. Low transmission noise through the band improves wearer acceptance. They can be comfortably stored around the neck when not in use, particularly convenient when moving in and out of noisy environments frequently. The band is washable and reusable and the 3M™ E-A-RCaps™ replacement pods are replaceable and available to purchase separately.