Position TrueBlot® magnetic separator, 6
Catalog # ROCKTMS-15-50
Supplier: Rockland Immunochemicals
Specifications
- Beskrivning:15/50 Tube TrueBlot® magnetic separator
- Shipping conditions:At ambient temperatures
- Förp.:1
Specifications
About this item
6x1,5 ml Position magnetic separator
- Magnetic separation
- Six 1,5 ml microcentrifuge tubes
A magnetic separation unit designed to accommodate up to six 1,5 ml microcentrifuge tubes in order to separate magnetic particles contained in those tubes. Optimal working volume is up to 1 ml.
Magnetic separator unit can be used to hold, and separate magnetic particles.