ContactUsLinkComponent
Replacement membrane filters, ali-Q™ 2
Replacement membrane filters, ali-Q™ 2
Katalog # VIST9020-4004
Leverantör:  VISTALAB TECHNOLOGIES
undefined
Replacement membrane filters, ali-Q™ 2
Katalog # VIST9020-4004
Leverantör:  VISTALAB TECHNOLOGIES
Specifikationer
  • Porstorlek:
    0.22 µm
  • Steril:
    -
  • Material:
    PTFE
  • Beskrivning:
    ali-Q™ replacement membrane filter
  • Seq:
    02
  • Förp.:
    5