Anti-ALB rabbit polyclonal antibody
Katalog # NOVUNBP1-32458
Leverantör: Novus Biologicals
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Albumin
- Antigensymbol:ALB
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Gen-ID:213
- Antigen synonymer:growth-inhibiting protein 20|PRO1341|DKFZp779N1935|serum albumin|PRO0883|cell growth inhibiting protein 42|albumin|PRO0903
- Storage buffer:PBS (pH 7,0), 1,0% BSA and 20% Glycerol
- Förvaringstemperatur:Aliquot and store at −20 or −80 °C. Avoid freeze-thaw cycles.
- Koncentration:0,38 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the C-terminus region of human Albumin. The exact sequence is proprietary.
- Rening:Immunogen affinity purified
- Förp.:0,1 mL
Specifikationer
Om denna produkt
The Albumin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Albumin. This antibody reacts with human, mouse, rat. The Albumin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: Albumin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat