Anti-NLRP1/NALP1 Rabbit polyclonal antibody
Katalog # NOVUNB100-56148SS
Leverantör: Novus Biologicals
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:NLRP1/NALP1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- Immunutfällning:Yes
- Isotyp:IgG
- Reaktivitet:Human
- Western blot:Yes
- Gen-ID:22861
- Antigen synonymer:Nucleotide-binding domain and caspase recruitment domain|NACHT|Caspase recruitment domain-containing protein 7|LRR and PYD domains-containing protein 1|CARD7SLEV1|NALP1caspase recruitment domain protein 7|nucleotide-binding oligomerization domain|NLR family|NACPP1044|pyrin domain containing 1|leucine rich repeat and pyrin domaincontaining 1|KIAA0926DEFCAP-L/S|DEFCAPVAMAS1|CLR17.1|Death effector filament-forming ced-4-like apoptosis protein|leucine rich repeat and PYD (pyrin domain) containing 1|DKFZp586O1822|LRR and PYD containing protein 1|leucine rich repeat and PYD containing 1
- Storage buffer:Whole antisera
- Förvaringstemperatur:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:A synthetic peptide corresponding to amino acids 161-180 (PSSPDHESPSQESPNAPTST) of human NALP1/NAC/CARD7 was used as immunogen, GenBank no. NP_127497.1.
- Rening:Unpurified
- Förp.:25 µl
Specifikationer
Om denna produkt
The NLRP1 / NALP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to NLRP1 / NALP1. This antibody reacts with human. The NLRP1 / NALP1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: NLRP1/NALP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human