Powered air respirator back pack adapters, Versaflo™
Catalog # MMMATR-655
Supplier: 3M
Specifications
- Beskrivning:Back pack adapter
- Förp.:1
Specifications
About this item
The Versaflo™ Backpack Adaptor TR-655 allows users to wear Versaflo™ Powered Air Turbo TR-600 or TR-800 unit on their back when used with Versaflo™ Backpack Harness BPK-01.
- Effective alternative to wearing the powered air turbo on a belt
- Designed for use alongside Versaflo™ Backpack Harness BPK-01
This adaptor is specially designed for use with Versaflo™ Powered Air Turbo and Versaflo™ Backpack Harness BPK-01. It offers users the versatility to choose how to carry their powered air turbo according to the task they’re completing.