Orders LinkContactUsLinkComponent
VWR®, Advanced hotplates
VWR®, Advanced hotplates
Hotplate, ceramic plate, 180×180 mm
Katalog # 442-1269
Leverantör:   VWR Collection
VWR®, Advanced hotplates
Katalog # 442-1269
Leverantör:  VWR Collection
VWR®, Advanced hotplates
VWR®, Advanced hotplates
Hotplate, ceramic plate, 180×180 mm
Price
9 350,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
VWR® Advanced hotplates are designed to deliver accurate and repeatable results in all research, academic and industrial applications. With temperature ranges up to 500 °C VWR hotplates provide superior temperature control.
  • Excellent temperature uniformity
  • Easy to read, large LCD displays temperatures
  • Industry-leading safety features
  • Cool touch and chemical resistant housing
  • 5 year warranty
These microprocessor-controlled units feature a front facing control panel with an easy to read LCD that displays temperature settings and intuitive icons.
Reflective, white ceramic top is chemical-resistant and easy to clean. A built-in memory recall provides users with the last set temperature, even after the unit has been turned off. For additional safety, a large, hot symbol warning light is illuminated when the hot plate is in use, this remains lit until the top plate is below 40 °C. Units are equipped with redundant safety controls, which will shut down unit in case of heater malfunction.
The low profile design utilises minimal bench space and allows use within a fume hood. Unit housing is constructed of a heat and chemical resistant polymer that remains cool to the touch during operation. The spill resistant design channels fluids away from internal components in the event of a spill.
Corresponding magnetic hotplate stirrers (heating and stirring 442-1265, 442-1268, 442-1270 and 442-1274) or magnetic stirrers (stirring only 442-1310) are also available.
Certifications: RoHS compliant. TUV listed.
Delivery information: Units are supplied with a 234 cm three-wire cord and plug.
Specifikationer
  • Beskrivning:
    Hotplate, ceramic plate, 180×180 mm
  • Seq:
    001
  • Cat. No.:
    442-1269
  • Modell:
    442-1269
  • Plattmaterial:
    Glaskeramik
  • Vikt:
    2,8 kg
  • B×D×H:
    224×307×122 mm
  • Temperaturområde:
    Ambient +5...500 °C
  • Temperaturstabilitet:
    Top plate: ±3% >100 °C; ±2% ≤100 °C
    with temperature probe: ±1% >100 °C; ±1% ≤100 °C
  • Plattans mått:
    180×180 mm