Precision Digital ProtEX Explosion-Proof Process Meters
Process meter, explosion-proof, input loop powered with backlight
Katalog # MFLX30004-68
CE Compliant
Leverantör: Avantor Fluid Handling
Precision Digital ProtEX Explosion-Proof Process Meters
Katalog # MFLX30004-68
Leverantör: Avantor Fluid Handling
CE Compliant
Gå till:
Product Details & Documents
Four sensors operate as through-glass buttons for easy on-site programming without removing the cover and exposing the electronics.
- High-contrast backlit LCD is easy to read from a distance and under most lighting conditions
- 5-digit rate and 7-character total/tag display
- 32-point, square root, or exponential linearization
- 90° rotatable display
- 5-digit password prevents unauthorized changes to the programmed parameter settings
- Explosion-proof, IP68, NEMA 4X enclosure
- FM, CSA, ATEX, IECEx, CE approved
The ProtEX PD6800 is a rugged, explosion-proof process meter designed for demanding applications in hazardous areas or in harsh environmental conditions. The meter derives all of its power from the 4 to 20 mA loop. Easily program using the four SafeTouch™ through-glass buttons without having to remove the cover or use the four mechanical buttons. Unit features backlit display, min/max readings, password protection, and alarm indication. Die-cast aluminum enclosure has two ¾" threaded conduit holes and integrated pipe or wall mounting flanges.
Specifikationer
- Beskrivning:Process meter, explosion-proof, input loop powered with backlight
- Cat. No.:MFLX30004-68
- Garanti:3 years