CPC® Quick-Disconnect Fittings, Threaded Inserts
Om denna produkt
No more struggling - just press thumb latch.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
10 Alternativ tillgängliga nedan
Produktdetaljer och dokument
No more struggling - just press thumb latch.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
Documents
Certificates
Sök efter LOT-nummerSpecifications for Product Family
Specifikationer
- Catalog No:
- MFLX06361-81BLK
- MFLX06361-91BLK
- MFLX06361-29BLK
- MFLX06361-31BLK
- MFLX06361-65BLK
- MFLX06361-29
- MFLX06361-31
- MFLX06361-65
- MFLX06361-91
- MFLX06361-81
- Max. operating pressure:
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- Max. temperature:
- 71 °C (160 °F)
- 71 °C (160 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- 71 °C (160 °F)
- 71 °C (160 °F)
- Max. vacuum:
- 711 mm Hg (28" Hg)
- 711 mm Hg (28" Hg)
- 711 mm Hg (28" Hg)
- 711 mm Hg (28" Hg)
- 711 mm Hg (28" Hg)
- 711 mm Hg (28" Hg)
- 711 mm Hg (28" Hg)
- 711 mm Hg (28" Hg)
- 711 mm Hg (28" Hg)
- 711 mm Hg (28" Hg)
- Seal material:
- EPDM
- EPDM
- Buna N
- Buna N
- Buna N
- Buna N
- Buna N
- Buna N
- EPDM
- EPDM
- Sterilisering:
- Gamma
- Gamma
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Gamma
- Gamma
- Valve spring material:
- 316 Stainless Steel
- 316 Stainless Steel
- 316 Stainless Steel
- 316 Stainless Steel
- 316 Stainless Steel
- 316 Stainless Steel
- 316 Stainless Steel
- 316 Stainless Steel
- Garanti:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Specifikationer
Product Family Options
Bulk product
Produktinformation
- TypAnslutningstypFitting materialFlow patternQuick-disconnect flow sizeThread size NPT(M) (port 1)Thread size NPT(M) (port 2)Supplier No.TillgänglighetPrice
- MFLX06361-81BLKQuick-Disconnect Fitting, Thread Insert, Polypropylene, Straight-Through; ¹/₄" ID, PLC2400412; 25/PKQuick-Disconnect Fitting, Thread Insert, Polypropylene, Straight-Through; ¹/₄" ID, PLC2400412; 25/PKQuick-Disconnect to ThreadPolypropyleneStraight¼"¼"¼"PLC2400412 (25PK)Log in för att kolla tillgänglighet och ert pris1 * 25 Items2 050,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-81BLKMax. operating pressure120 psiMax. temperature71 °C (160 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialEPDMSteriliseringGammaValve spring materialGaranti1 year - MFLX06361-91BLKQuick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, ¹/₄" ID, PLCD2400412; 25/PKQuick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, ¹/₄" ID, PLCD2400412; 25/PKValvedQuick-Disconnect to ThreadPolypropyleneStraight¼"¼"¼"PLCD2400412 (25PK)Log in för att kolla tillgänglighet och ert pris1 * 25 Items3 475,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-91BLKMax. operating pressure120 psiMax. temperature71 °C (160 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialEPDMSteriliseringGammaValve spring material316 Stainless SteelGaranti1 year - MFLX06361-29BLKQuick-Disconnect Fitting, Threaded Insert, Acetal, Valved, ¹/₈" ID, PMCD2402; 25/PKQuick-Disconnect Fitting, Threaded Insert, Acetal, Valved, ¹/₈" ID, PMCD2402; 25/PKValvedQuick-Disconnect to ThreadAcetalStraight⅛"⅛"⅛"PMCD2402 (25PK)Log in för att kolla tillgänglighet och ert pris1 * 25 Items2 325,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-29BLKMax. operating pressure120 psiMax. temperature82 °C (180 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialBuna NSteriliseringAutoclave or ethylene oxide (ETO)Valve spring material316 Stainless SteelGaranti1 year - MFLX06361-31BLKQuick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₈" Flow Size; ¹/₄" ID, PMCD2404; 25/PKQuick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₈" Flow Size; ¹/₄" ID, PMCD2404; 25/PKValvedQuick-Disconnect to ThreadAcetalStraight⅛"¼"⅛"PMCD2404 (25PK)Log in för att kolla tillgänglighet och ert pris1 * 25 Items2 375,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-31BLKMax. operating pressure120 psiMax. temperature82 °C (180 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialBuna NSteriliseringAutoclave or ethylene oxide (ETO)Valve spring material316 Stainless SteelGaranti1 year - MFLX06361-65BLKQuick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₄" Flow Size; ¹/₄" ID, PLCD24004; 25/PKQuick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₄" Flow Size; ¹/₄" ID, PLCD24004; 25/PKValvedQuick-Disconnect to ThreadAcetalStraight¼"¼"¼"PLCD24004 (25PK)Log in för att kolla tillgänglighet och ert pris1 * 25 Items2 225,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-65BLKMax. operating pressure120 psiMax. temperature82 °C (180 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialBuna NSteriliseringAutoclave or ethylene oxide (ETO)Valve spring material316 Stainless SteelGaranti1 year
Down pack product
Produktinformation
- TypAnslutningstypFitting materialFlow patternQuick-disconnect flow sizeThread size NPT(M) (port 1)Thread size NPT(M) (port 2)Supplier No.TillgänglighetPrice
- MFLX06361-29Quick-Disconnect Fitting, Threaded Insert, Acetal, Valved, ¹/₈" ID, PMCD2402; 1/PKQuick-Disconnect Fitting, Threaded Insert, Acetal, Valved, ¹/₈" ID, PMCD2402; 1/PKValvedQuick-Disconnect to ThreadAcetalStraight⅛"⅛"⅛"Log in för att kolla tillgänglighet och ert pris1 * 1 Items155,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-29Max. operating pressure120 psiMax. temperature82 °C (180 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialBuna NSteriliseringAutoclave or ethylene oxide (ETO)Valve spring material316 Stainless SteelGaranti1 year - MFLX06361-31Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₈" Flow Size; ¹/₄" ID, PMCD2404; 1/PKQuick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₈" Flow Size; ¹/₄" ID, PMCD2404; 1/PKValvedQuick-Disconnect to ThreadAcetalStraight⅛"¼"⅛"Log in för att kolla tillgänglighet och ert pris1 * 1 Items155,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-31Max. operating pressure120 psiMax. temperature82 °C (180 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialBuna NSteriliseringAutoclave or ethylene oxide (ETO)Valve spring material316 Stainless SteelGaranti1 year - MFLX06361-65Quick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₄" Flow Size; ¹/₄" ID, PLCD24004; 1/PKQuick-Disconnect Fitting, Thread Insert, Acetal, Valved, ¹/₄" Flow Size; ¹/₄" ID, PLCD24004; 1/PKValvedQuick-Disconnect to ThreadAcetalStraight¼"¼"¼"Log in för att kolla tillgänglighet och ert pris1 * 1 Items160,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-65Max. operating pressure120 psiMax. temperature82 °C (180 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialBuna NSteriliseringAutoclave or ethylene oxide (ETO)Valve spring material316 Stainless SteelGaranti1 year - MFLX06361-91Quick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, ¹/₄" ID, PLCD2400412; 1/PKQuick-Disconnect Fitting, Threaded Insert, Polypropylene, Valved, ¹/₄" ID, PLCD2400412; 1/PKValvedQuick-Disconnect to ThreadPolypropyleneStraight¼"¼"¼"Log in för att kolla tillgänglighet och ert pris1 * 1 Items235,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-91Max. operating pressure120 psiMax. temperature71 °C (160 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialEPDMSteriliseringGammaValve spring material316 Stainless SteelGaranti1 year - MFLX06361-81Quick-Disconnect Fitting, Thread Insert, Polypropylene, Straight-Through; ¹/₄" ID, PLC2400412; 1/PKQuick-Disconnect Fitting, Thread Insert, Polypropylene, Straight-Through; ¹/₄" ID, PLC2400412; 1/PKQuick-Disconnect to ThreadPolypropyleneStraight¼"¼"¼"Log in för att kolla tillgänglighet och ert pris1 * 1 Items130,00 krSpecifikationer:
More
Specifications
Cat. No.MFLX06361-81Max. operating pressure120 psiMax. temperature71 °C (160 °F)Max. vacuum711 mm Hg (28" Hg)Seal materialEPDMSteriliseringGammaValve spring materialGaranti1 year