Heavy-Duty Tubing Pinch Clamp, Polysulfone
Katalog # MFLX20001-00
Leverantör: Avantor Fluid Handling
Specifikationer
- Beskrivning:Heavy-Duty Tubing Pinch Clamp, Polysulfone
- Material:Polysulfon
- Garanti:3 Months
- Förp.:1
Specifikationer
Om denna produkt
Durable design handles higher working pressures.
- Quick installation with positive swing-away locking screw
- Smooth contours allow for easy cleaning
Use this clamp with tubing or hose - it will meet the working pressure of either. High-strength polysulfone closure provides excellent durability, can be continually reused, and resists most chemicals. Simply align the hose within the clamp; mating surfaces on the clamp indicate complete closure. Typical applications include sterile filling, media bag connections, pumping, liquid control and high-purity fluid transfers. FDA-compliant polysulfone material can be sterilized by autoclave or gamma radiation.
- Beskrivning:Heavy-Duty Tubing Pinch Clamp, Polysulfone
- Material:Polysulfon
- Environmentally Preferable:
- Garanti:3 Months
- Förp.:1