Pre-sterilized individually wrapped blue pestle, Axygen®
Katalog # AXYGPES-15-B-SI
Leverantör: Corning
Specifikationer
- För:1.5 - 2.0 ml
- Beskrivning:Pre-sterilized, individually wrapped pestle
- Förp.:400
Specifikationer
Om denna produkt
Durable and inert polypropylene construction
- Designed to fit 1.5 and 2.0 ml centrifuge tubes
- Individually wrapped and sterilized