Agarose LE, molecular biology grade, Axygen®
Leverantör: Corning
Certifikat
Om denna produkt
Axygen® Agarose Low Electroendosmosis (LE) is a high quality molecular biology grade agarose suitable for analytical and preparative electrophoresis of nucleic acids.
Separation range: 1 to 2,5 Kbp.
Specifikationer
- Catalog No:
- AXYGAGR-LE-1001
- AXYGAGR-LE-5001
- Environmentally Preferable:
- Low Manufacturing Impact
- Geleringstemperatur:
- 36 °C (±1,5 °C) [1,5%]
- 36 °C (±1,5 °C) [1,5%]
- Smältpunkt:
- 88°C (±1,5°C) [1,5%]
- 88°C (±1,5°C) [1,5%]
- Miljöcertificering:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa