Belt bag, Force®8
Katalog # JSPABPT170-011-000
Leverantör: JSP
Specifikationer
- Beskrivning:Force®8 Belt bag, for masks and filters
- Förp.:10
Specifikationer
Om denna produkt
Force®8 Belt Bag holds mask and filters when not in use, keeping them free from dirt and damage.
- Convenient accessibility for Force®8 half-masks
- Filters and mask remain clean and safe when in bag
- Increased lifespan for stored items
This robust bag is secured with a wide, comfortable strap and string clip. The main compartment allows for the storage of the Force®8 half-mask without the filters attached. On either side of this main compartment are two more compartments suitable for a filter each. This keeps a complete unit safe and clean.