Safety helmet, uvex pheos planet B-S-WR
Catalog # 112-6185
Supplier: UVEX
Specifications
- Färg:Vit
- Beskrivning:Safety helmet with ventilation and shortened brim
- Environmentally Preferable:Sustainable Materials,Low Manufacturing Impact,Product Transparency and Disclosure,Sustainable Packaging,Waste Reducing
- Modell:uvex pheos planet B-S-WR
- Förp.:1
Specifications
About this item
Lightweight helmets made from HDPE.
- Suspension harness with wheel ratchet system for variable width adjustment
- Safety helmet with helmet torch slot on the front
- Euroslot adapters on the side (30 mm) for attaching earmuffs and the uvex pheos visor system
- The shortened brim ensures unrestricted upwards vision
- Six-point textile band attachment ensures excellent fit and comfort
- Three adjustable air vents for maximum ventilation
Ideal for industry, road construction works, sewage and pipeline construction, concrete/construction work, brickwork, stonemasonry, plasterwork, demolition/dismantling work, bridge/tunnel construction, high street construction and metalwork.
Adjustable size range: 520 to 610 mm
Certifications: Meets EN 397 standard and additional requirements for very low temperatures (−30 °C) and molten metal (MM) splash.