Waste Tube Connection, b.safe
Catalog # BOHLR1105-25
Supplier: Bohlender
Specifications
- Gänga:2× GL 25 (male)
- Beskrivning:Cabinet mount, usable length 110 mm, for bore Ø 28 mm
- Förp.:1
Specifications
About this item
For safe connection of canisters in safety cabinets to a waste disposal system with b.safe Waste Tubes.
- Made of static dissipative PE-EX
- A suitable attachment for fire protection is available separately
Mount for safety cabinets with a wall thickness of max. 100 mm and bore Ø 28 mm, lock nut with silicone gaskets for clamp fastening, 2 connections GL 25 male for connection to b.safe Waste Tubes.