ContactUsLinkComponent
Sealing ring for waste pipes, b.safe
Sealing ring for waste pipes, b.safe
Katalog # BOHLR1161-01
Leverantör:  Bohlender
undefined
Sealing ring for waste pipes, b.safe
Katalog # BOHLR1161-01
Leverantör:  Bohlender
Specifikationer
  • Beskrivning:
    Sealing ring for waste pipes tube, PTFE, Ext.Ø 32 mm
  • Förp.:
    1
Specifikationer
Om denna produkt
Conical sealing ring made of static dissipative PTFE-EX.
Suitable spare part for clamp connections for tube Ext.Ø 32 mm.