Blind cap for waste pipes, b.safe
BOHLR1160-05
- Beskrivning:Blind cap for waste pipes, PE, for thread M38×2
- Förp.:1
Cap made of PE-EX with pressed-in conical sealing plate made of static-dissipative PTFE-EX.
Suitable sealing cap for clamp connections for tube Ext.Ø 32 mm.