Grade 2589 D filter papers for technical use, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
A very thick filter with high wet strength. Offers medium to slow flow rate and retains very fine precipitates.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.
Specifications
- Catalog No:
- WHAT10343978
- 512-0411
- Klass:
- 2589 d
- 2589 d
- Vikt:
- 500 g/m²
- 500 g/m²
- Tjocklek:
- 1000 µm
- 1000 µm
- Partikelretention:
- 2 - 6 µm
- 2–6 µm
- Filtreringshastighet:
- 235 s (Herzberg)
- 235 s (Herzberg)