Orders LinkContactUsLinkComponent
Nitrile gloves, Kimtech™ Opal™
Nitrile gloves, Kimtech™ Opal™
   Hållbara alternativ tillgängliga
Leverantör:   Ansell
Nitrile gloves, Kimtech™ Opal™
Leverantör:  Ansell
   Hållbara alternativ tillgängliga
Certifikat
Nitrile gloves, Kimtech™ Opal™
5 Alternativ tillgängliga nedan
Produktdetaljer och dokument
Kimtech™ Opal™ nitrile gloves guard against contamination by chemical splash and microorganism hazards. Powder-free, ambidextrous, with beaded cuff and textured fingertips. The absence of natural rubber latex and vulcanising agents reduces the risk of TYPE I and IV glove associated skin reactions.
  • Assured compliance
  • Formulated without thiurams, thiazoles, guanidines, carbamates, and no sulphur added
  • Soft, comfortable, and easy to don
  • Textured fingertips enhance grip and tactile sensitivity for safer and more efficient processes
  • Dark colour to enhance concentration on task
  • Available in a wide range of sizes, from XS to XL
These gloves guard against contamination by chemical splash and microorganism hazards, delivering seamless protection when and where it counts. Powder-free gloves that are ideal for use in research and production facilities, life sciences, and non sterile drug manufacturing applications. The gloves feature an innovative approach using a thin nitrile polymer with a tip thickness of just 0,07 mm, but with excellent tensile strength suitable for rigorous process use. Easy to don, soft and comfortable, they are suitable for extended wearing time.
The absence of natural rubber latex, sulphur and vulcanising agents reduces the risk of TYPE I and TYPE IV glove associated skin reactions, protecting the wearer as well as the application. Precision manufacturing reduces the risk of contamination and ensures the nitrile gloves offer high levels of regulatory compliance. Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials, and the ambidextrous gloves have been extensively tested under stringent conditions. In addition, the thin nitrile construction and efficient packaging mean that the chemical resistant gloves can reduce waste by up to 50%.
Certifications: EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS), Cat. III; AQL 1,5
Food contact approved
Dokument
Specifikationer för produkt
Specifikationer
  • Catalog No:
  • 112-6223
  • 112-6224
  • 112-6225
  • 112-6226
  • 112-6227
  • Ej handspecifika/handspecifika:
  • Ej handspecifika
  • Ej handspecifika
  • Ej handspecifika
  • Ej handspecifika
  • Ej handspecifika
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Engångs/Återanvändbar:
  • Engångs
  • Engångs
  • Engångs
  • Engångs
  • Engångs
  • Latex/latexfri:
  • Latexfria
  • Latexfria
  • Latexfria
  • Latexfria
  • Latexfria
  • Pudrad/puderfri:
  • Puderfria
  • Puderfria
  • Puderfria
  • Puderfria
  • Puderfria
  • Material:
  • Nitril
  • Nitril
  • Nitril
  • Nitril
  • Nitril
  • Yta:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Kragmodell:
  • Med rullad kant
  • Med rullad kant
  • Med rullad kant
  • Med rullad kant
  • Med rullad kant
  • EN standard:
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
  • Godkända för livsmedelskontakt:
  • +
  • +
  • +
  • +
  • +
Specifikationer
Product Family Options
Produktinformation
  • Handskstorlek
    Färg
    Längd
    Förp.
    Tillgänglighet
    Pris
  • XS
    Mörkblå
    240 mm
    200/box
    Log in för tillgänglighet
    Log in för tillgänglighet
    1 * 200 Items
    1 * 200 Items - 460,00 kr
    460,00 kr
    1 * 200 Items
       460,00 kr
    Specifikationer:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6223
    Ej handspecifika/handspecifika
    Ej handspecifika
    Environmentally Preferable
    Waste Reducing
    Engångs/Återanvändbar
    Engångs
    Latex/latexfri
    Latexfria
    Pudrad/puderfri
    Puderfria
    Material
    Nitril
    Yta
    Textured fingertips
    Kragmodell
    Med rullad kant
    EN standard
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Godkända för livsmedelskontakt
    +
      Waste Reducing
  • S
    Mörkblå
    240 mm
    200/box
    Log in för tillgänglighet
    Log in för tillgänglighet
    1 * 200 Items
    1 * 200 Items - 460,00 kr
    460,00 kr
    1 * 200 Items
       460,00 kr
    Specifikationer:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6224
    Ej handspecifika/handspecifika
    Ej handspecifika
    Environmentally Preferable
    Waste Reducing
    Engångs/Återanvändbar
    Engångs
    Latex/latexfri
    Latexfria
    Pudrad/puderfri
    Puderfria
    Material
    Nitril
    Yta
    Textured fingertips
    Kragmodell
    Med rullad kant
    EN standard
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Godkända för livsmedelskontakt
    +
      Waste Reducing
  • M
    Mörkblå
    240 mm
    200/box
    Log in för tillgänglighet
    Log in för tillgänglighet
    1 * 200 Items
    1 * 200 Items - 460,00 kr
    460,00 kr
    1 * 200 Items
       460,00 kr
    Specifikationer:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6225
    Ej handspecifika/handspecifika
    Ej handspecifika
    Environmentally Preferable
    Waste Reducing
    Engångs/Återanvändbar
    Engångs
    Latex/latexfri
    Latexfria
    Pudrad/puderfri
    Puderfria
    Material
    Nitril
    Yta
    Textured fingertips
    Kragmodell
    Med rullad kant
    EN standard
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Godkända för livsmedelskontakt
    +
      Waste Reducing
  • L
    Mörkblå
    240 mm
    200/box
    Log in för tillgänglighet
    Log in för tillgänglighet
    1 * 200 Items
    1 * 200 Items - 480,00 kr
    480,00 kr
    1 * 200 Items
       480,00 kr
    Specifikationer:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6226
    Ej handspecifika/handspecifika
    Ej handspecifika
    Environmentally Preferable
    Waste Reducing
    Engångs/Återanvändbar
    Engångs
    Latex/latexfri
    Latexfria
    Pudrad/puderfri
    Puderfria
    Material
    Nitril
    Yta
    Textured fingertips
    Kragmodell
    Med rullad kant
    EN standard
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Godkända för livsmedelskontakt
    +
      Waste Reducing
  • XL
    Mörkblå
    240 mm
    170/box
    Log in för tillgänglighet
    Log in för tillgänglighet
    1 * 170 Items
    1 * 170 Items - 390,00 kr
    390,00 kr
    1 * 170 Items
       390,00 kr
    Specifikationer:
      Waste Reducing
    More
    Specifications
    Cat. No.
    112-6227
    Ej handspecifika/handspecifika
    Ej handspecifika
    Environmentally Preferable
    Waste Reducing
    Engångs/Återanvändbar
    Engångs
    Latex/latexfri
    Latexfria
    Pudrad/puderfri
    Puderfria
    Material
    Nitril
    Yta
    Textured fingertips
    Kragmodell
    Med rullad kant
    EN standard
    EN ISO 374-1 Type B (KPT), EN 374-4, EN ISO 374-5 (VIRUS)
    Godkända för livsmedelskontakt
    +
      Waste Reducing