Washroom cleaner and descaler, SURE®
Katalog # 115-3070
Leverantör: Diversey
Specifikationer
- Beskrivning:Washroom cleaner and descaler, 6×1 L
- Seq:01
- Environmentally Preferable:Sustainable Materials,Safer Human and Environmental Health,Sustainable Packaging
- Förp.:6 Bottle
Specifikationer
Om denna produkt
Concentrated, highly effective product for daily and periodical cleaning of water resistant hard surfaces in washrooms.
- Concentrated formulation gives excellent economy of use
- Effective cleaning and descaling even in hard water areas
- Suitable for use on intact chrome and stainless steel surfaces
- Cleans without leaving residue
- No fragrance or dye
- Contains plant-based ingredients from renewable sources
- Decomposed by natural processes
- No environmental hazard classifications, no safety warnings for product in use for user
SURE® washroom cleaner and descaler is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: OECD 301B test.
EU-Ecolabel (EU: NL/020/045).
Detergent Regulation, EC 648/2004.
EU-Ecolabel (EU: NL/020/045).
Detergent Regulation, EC 648/2004.