Orders LinkContactUsLinkComponent
Scissors, duck, Rubis®
Scissors, duck, Rubis®
Straight, blunt/blunt, 92 mm, 1 mm
Katalog # 233-0224
Leverantör:   OUTILS RUBIS
Scissors, duck, Rubis®
Katalog # 233-0224
Leverantör:  OUTILS RUBIS
Scissors, duck, Rubis®
Scissors, duck, Rubis®
Straight, blunt/blunt, 92 mm, 1 mm
Price
530,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Micro scissors with beak-shaped tips.
  • Produced from hardened stainless steel AISI420
  • Highly resistant against acids and alkaline
  • With ergonomically designed Rubis Twist® grips
  • No Medical Device (MD) product
Made in Switzerland to the highest quality standards. Rubis products are the result of traditional craftsmanship, strict throughput controls and a finishing process under the magnifying glass. No tool leaves the factory without having passed the strict quality controls. Neither scratches nor inaccuracies in material or surface are permitted.
Specifikationer
  • Typ:
    Straight, blunt/blunt
  • Längd:
    92 mm
  • Tip size:
    1 mm