Cut resistant gloves, ActivArmr® 70-761
Supplier: Ansell
Certificates
About this item
These gloves made from Kevlar®, nylon and acrylic fibres provide the optimal combination of cut resistance, comfort and dexterity.
- Reinforced thumb crotch with 100% Kevlar®
- High level of cut protection
- Protection from heat and light molten splashes
- Excellent abrasion resistance due to exclusive fibre blend
- Design to fit the natural contours of the hand
Ideal for assembling metal parts and components, handling sharp or irregular-edged metal parts, protection from heat & light molten splashes, welding and metal finish inspection.
Certifications: EN 388:2016 (1X4XD), EN 407 (X1XXXX); Cat. III
Specifications
- Catalog No:
- 112-3739
- 112-3738
- 112-3740
- 112-3741
- 112-3737
- 112-3742
- Modell:
- 70-761
- 70-761
- 70-761
- 70-761
- 70-761
- 70-761
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Fodermaterial:
- Nylon/Kevlar®
- Nylon/Kevlar®
- Nylon/Kevlar®
- Nylon/Kevlar®
- Nylon/Kevlar®
- Nylon/Kevlar®
- Mätare:
- 10
- 10
- 10
- 10
- 10
- 10
- Kragmodell:
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- EN standard:
- EN 388:2016 (1X4XD), EN 407 (X1XXXX)
- EN 388:2016 (1X4XD), EN 407 (X1XXXX)
- EN 388:2016 (1X4XD), EN 407 (X1XXXX)
- EN 388:2016 (1X4XD), EN 407 (X1XXXX)
- EN 388:2016 (1X4XD), EN 407 (X1XXXX)
- EN 388:2016 (1X4XD), EN 407 (X1XXXX)