General purpose gloves, Perfect Poly® Skin
Leverantör: HONEYWELL SAFETY
Certifikat
Om denna produkt
Polyamide with its polyurethane coating provide excellent abrasion and tear resistance. Non pilling, the polyamide is suitable for precision work. The ventilated back and ultra light coating optimise the glove's breathability.
- Offers maximum comfort and added sturdiness
- The knitting technique helps to obtain a glove that conforms snugly to the hand as well as outstanding dexterity
- The ultra tight weave gives the glove perfect support, limits the penetration of dirt and offers a good touch
- Certified as 'silicone-free' to ensure their use in painting
- Its blue colour and grey coating make the glove more soilproof, thus ensuring a longer life
Ideal for the handling of fine products in a dry or slightly greasy or soiled environment.
Gauge: 15
Colour: blue/grey
EN 388 (2121), Cat. III
Certifications: EN 388:2016 (2110X); Cat. II
Specifikationer
- Catalog No:
- 112-1651
- 112-1650
- 112-1649
- 112-1648
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Fodermaterial:
- Polyamid
- Polyamid
- Polyamid
- Polyamid
- Yta:
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Mätare:
- 18
- 18
- 18
- 18
- Kragmodell:
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- EN standard:
- EN 388:2016 (2110X)
- EN 388:2016 (2110X)
- EN 388:2016 (2110X)
- EN 388:2016 (2110X)