Affinity protein expression and purification systems
Calmodulin affinity resin, Calmodulin binding peptide (CBP) affinity resin, 10 mL
Catalog # AGLS214303
Supplier: AGILENT
Affinity protein expression and purification systems
Catalog # AGLS214303
Supplier: AGILENT
CAS Number:
Jump to:
Product Details & Documents
The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
- pCAL vectors derived from T7 RNA polymerase-based pET vectors
- Novel purification tag combines small size, gentle elution and high-affinity binding
- Easy tag removal resulting in native protein
Delivery information: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
- Förp.:10 mL
- Beskrivning:Calmodulin affinity resin
- Details:Calmodulin binding peptide (CBP) affinity resin