VWR®, L-Glutamine
Katalog # 392-0441
Leverantör: VWR Chemicals
CAS-nummer:
Specifikationer
- Medieformat:Liquid
- Förp.:100 mL
- Beskrivning:L-Glutamine, 100X, 200 mM
Specifikationer
Om denna produkt
L-Glutamine is an essential amino acid required by virtually all mammalian and insect cells grown in culture. It is a crucial component of many cell culture media and serves as a major energy source for cells in culture.