VWR®, Cell culture media, Minimum Essential Medium (MEM)
Leverantör: VWR Chemicals
Certifikat
Om denna produkt
Minimum Essential Medium (MEM) with Earle’s Balanced Salts is a modification of Eagle’s earlier Basal Medium (BME) which contains a higher concentration of essential nutrients. These media promote the growth of a variety of normal and transformed cells. Since they contain Earle’s Balanced Salts, they are suitable for use in atmospheres charged with CO₂ gas.
Specifikationer
- Kalcium:Yes
- Magnesium:Yes
- HEPES:No
- Sodium bicarbonate:Yes
- Sodium pyruvate:No
- Phenol red:Yes
- Stable glutamine:No
- L-Glutamine:No
- Media Base:MEM/EBSS - Minimum Essential Medium with Earle's Balanced Salts
- Art.nr:392-0424
Specifikationer
Relaterad information
Certifikat
Sök efter LOT-nummerAvantor Cell Culture Solutions.pdf VWR Cell Culture Media