Murashige shoot tip rooting medium
Murashige shoot tip rooting medium 10×1 L, 10
Katalog # APOSPMM527-10X1L
Leverantör: Apollo Scientific
Murashige shoot tip rooting medium
Katalog # APOSPMM527-10X1L
Leverantör: Apollo Scientific
CAS-nummer:
Gå till:
Product Details & Documents
Contains macro/micronutrients, sucrose, agar andorganics as described by Malmgren, 1996 Plant tissue culture testedFull specification given in appendix.
- MFCD00243110
Specifikationer
Specifikationer
- Förp.:10
- Beskrivning:Murashige shoot tip rooting medium 10×1 L