AP-conjugate stabiliser, SeramunStab® STAP
Catalog # APOSBIL3116-100ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förp.:100 mL
- Beskrivning:SeramunStab® STAP AP-Conjugate stabiliser
- Seq:01
Specifications
About this item
Liquid product colourless odourless pH-value 5,9 to 6,1, Stabiliser for very low concentrated AP conjugates. Suitable for proteins resistant to pH values <6,0.