Restriction enzymes, Dpn I restriction enzymes
Catalog # AGLS500402
Leverantör: AGILENT
CAS-nummer:
Specifikationer
- Förp.:1
- Beskrivning:Dpn I
- Units:200 units
Specifikationer
Om denna produkt
Dpn I is an ideal enzyme for cloning and southern blot analysis.
- High purity enzyme preparation
Delivery: Includes 10X universal reaction buffer.
Caution: For research use only. Not for use in diagnostic procedures.