HRP-conjugate stabiliser, SeramunStab® Stplus
Catalog # APOSBIL3111-100ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förp.:100 mL
- Beskrivning:SeramunStab® Stplus HRP-conjugate stabiliser
- Seq:01
Specifications
About this item
Liquid product colourless to slightly yellow or greenish, odourless, pH 7,3 to 7,5. Stabiliser for conjugates with monomeric HRP.