Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteiner
Recombinant Protein G
Recombinant Protein G
Recombinant Protein G
Supplier:
Apollo Scientific
Certificates
Expand image
Recombinant Protein G
Supplier:
Apollo Scientific
About this item
Specifications
Protein/peptide type:
Recombinant
Protein/peptide name:
Protein G
Specifications