Order Entry
Sweden
ContactUsLinkComponent
Recombinant bacterial outer membrane protein-A
Recombinant bacterial outer membrane protein-A
Katalog # APOSBITP1617-1MG
Leverantör:  Apollo Scientific
CAS-nummer:  
Recombinant bacterial outer membrane protein-A
Katalog # APOSBITP1617-1MG
Leverantör:  Apollo Scientific
CAS-nummer:  

Specifikationer

  • Förp.:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    OmpA
  • Protein/peptide name:
    Bacterial Outer Membrane Protein-A

Specifikationer

Om denna produkt