Pepstatin A
Leverantör: Apollo Scientific
Certifikat
Om denna produkt
Pepstatin A is an inhibitor of acidic proteases.
Specifikationer
- Protein synonyms:Isovaleryl-L-val-L-val-statinyl-L-alastatine
- Protein/peptide name:Pepstatin A
Pepstatin A is an inhibitor of acidic proteases.