Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Peracetyl maltooctaose, anomeric mixture
Peracetyl maltooctaose, anomeric mixture
Peracetyl maltooctaose, anomeric mixture
Peracetyl maltooctaose, anomeric mixture
Supplier:
Apollo Scientific
Certificates
Expand image
Peracetyl maltooctaose, anomeric mixture
Supplier:
Apollo Scientific
About this item
:
Storage Temperature:
Omgivande