SpectraSynQ 1621
Catalog # APOSPC9725-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:160…220 °C/0,01mmHg
Density:1,8 g/cm³ (25 °C)
:GHS07
Storage Temperature:Omgivande
CAS Number:105809-15-2
MDL Number:MFCD08461619