1-Fluorohexane 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:373-14-8
:
Formula:C₆H₁₃F
MDL Number:MFCD00000454
:104.17 g/mol
Boiling Pt:92…93 °C
Density:0,8 g/cm³ (25 °C)
Flash Pt:27 °C
Storage Temperature:Omgivande
EINECS:206-763-8
UN:2929
ADR:6.1+3,II
:GHS02
:GHS07