2',4',6'-Trichloroacetanilide
Katalog # APOSPC3888-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:152…154 °C
:GHS07
Storage Temperature:Omgivande
CAS Number:363-40-6
Formula:C₈H₆Cl₃NO
MDL Number:MFCD02093332