2,3,6-Trifluoropyridine
Catalog # APOSPC10552-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:5 g
- Seq:3173
Specifications
About this item
:
:133.07 g/mol
Boiling Pt:100…102 °C
Density:1,499 g/cm³ (25 °C)
Flash Pt:30 °C
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Omgivande
CAS Number:3512-18-3
Formula:C₅H₂F₃N
MDL Number:MFCD03001158